CAS RN 3376-83-8
CAS RN 3376-83-8
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
(-)-Biotin Sulfoxide, TRC
CAS: 3376-83-8 Molecular Formula: C10 H16 N2 O4 S Molecular Weight (g/mol): 260.31 Synonym: 1H-Thieno[3,4-d]imidazole-4-pentanoic acid, hexahydro-2-oxo-, 5-oxide, [3aS-(3aα,4β,5β,6aα)]-,Biotin, 5-oxide (7CI),Biotin, 5-oxide, (-)- (8CI),Biotin, L-S-oxide (6CI),(-)-Biotin sulfoxide,Biotin l-sulfoxide,Biotin sulfoxide IUPAC Name: 5-[(3aS,4R,6aR)-2,5-dioxo-1,3,3a,4,6,6a-hexahydrothieno[3,4-d]imidazol-4-yl]pentanoic acid SMILES: OC(=O)CCCC[C@@H]1[C@H]2NC(=O)N[C@H]2CS1=O