accessibility menu, dialog, popup

UserName

CH3H3CCH3NO(E/Z)

CAS RN 3376-24-7

IUPAC Name: N-tert-butyl-1-phenylmethanimine oxide
Synonyms: phenyl T-butylnitrone
Molecular Weight (g/mol): 177.25
Molecular Formula: C11H15NO
InChi Key: IYSYLWYGCWTJSG-UHFFFAOYSA-N
SMILES: CC(C)(C)[N+]([O-])=CC1=CC=CC=C1
Molecular Weight (g/mol) 
  • (2)
  • (3)
Quantity 
  • (2)
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results