accessibility menu, dialog, popup

UserName

CAS RN 337508-62-0

IUPAC Name: 5-isothiocyanato-2,1,3-benzothiadiazole
Synonyms: 5-isothiocyanato-2,1,3-benzothiadiazole
Molecular Weight (g/mol): 193.24
Molecular Formula: C7H3N3S2
InChi Key: HHDWFDQYUFKNCO-UHFFFAOYSA-N
SMILES: S=C=NC1=CC2=NSN=C2C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results