accessibility menu, dialog, popup

UserName

PdCH3OOCH3OO

CAS RN 3375-31-3

IUPAC Name: palladium(2+) diacetate
Synonyms: palladium(II) acetate
Molecular Weight (g/mol): 224.51
Molecular Formula: C4H6O4Pd
InChi Key: YJVFFLUZDVXJQI-UHFFFAOYSA-L
SMILES: [Pd++].CC([O-])=O.CC([O-])=O
Quantity 
  • (4)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)
  • (1)
  • (1)
Percent Purity 
  • (3)
  • (2)
  • (2)
Physical Form 
  • (3)
  • (2)
Grade 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (4)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)
  • (1)
  • (1)
  • (3)
  • (2)
  • (2)
  • (3)
  • (2)
  • (2)