accessibility menu, dialog, popup

UserName

CAS RN 336628-67-2

FFFHOO

CAS RN 336628-67-2

IUPAC Name: 2-hydroxy-3-(trifluoromethyl)benzaldehyde
Synonyms: 2-hydroxy-3-(trifluoromethyl)benzaldehyde
Molecular Weight (g/mol): 190.12
Molecular Formula: C8H5F3O2
InChi Key: ZUNOXMJBNMKYOM-UHFFFAOYSA-N
SMILES: OC1=C(C=O)C=CC=C1C(F)(F)F
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results