accessibility menu, dialog, popup

UserName

CH3Br

CAS RN 3355-28-0

IUPAC Name: 1-bromobut-2-yne
Synonyms: 1-bromobut-2-yne
Molecular Weight (g/mol): 132.99
Molecular Formula: C4H5Br
InChi Key: LNNXOEHOXSYWLD-UHFFFAOYSA-N
SMILES: CC#CCBr
Molecular Weight (g/mol) 
  • (3)
  • (2)
  • (3)
Quantity 
  • (1)
  • (3)
  • (3)
  • (1)
Boiling Point 
  • (2)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (2)
  • (3)
  • (1)
  • (3)
  • (3)
  • (1)
  • (2)
  • (3)
  • (3)