accessibility menu, dialog, popup

UserName

CAS RN 33489-69-9

NOOONHF

CAS RN 33489-69-9

IUPAC Name: N-(4-fluorophenyl)-3-nitrobenzamide
Synonyms: N-(4-fluorophenyl)-3-nitrobenzamide
Molecular Weight (g/mol): 260.22
Molecular Formula: C13H9FN2O3
InChi Key: YPRHMRVPFAHCJX-UHFFFAOYSA-N
SMILES: [O-][N+](=O)C1=CC=CC(=C1)C(=O)NC1=CC=C(F)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results