CAS RN 333962-91-7
CAS RN 333962-91-7
IUPAC Name:
2-amino-6-{[(1H-imidazol-2-yl)methyl]sulfanyl}-4-(4-oxocyclohexa-2,5-dien-1-ylidene)-1,4-dihydropyridine-3,5-dicarbonitrile
Synonyms:
2-amino-6-[(1H-imidazol-2-ylmethyl)sulfanyl]-4-(4-oxocyclohexa-2,5-dien-1-ylidene)-1H-pyridine-3,5-dicarbonitrile
Molecular Weight (g/mol):
348.38
Molecular Formula:
C17H12N6OS
InChi Key:
LOLJUQOCQBYBGT-UHFFFAOYSA-N
SMILES:
NC1=C(C#N)C(=C2C=CC(=O)C=C2)C(C#N)=C(N1)SCC1=NC=CN1