accessibility menu, dialog, popup

UserName

CH3NCO

CAS RN 33375-06-3

IUPAC Name: (1-isocyanatoethyl)benzene
Synonyms: (1-isocyanatoethyl)benzene
Molecular Weight (g/mol): 147.18
Molecular Formula: C9H9NO
InChi Key: JJSCUXAFAJEQGB-UHFFFAOYNA-N
SMILES: CC(N=C=O)C1=CC=CC=C1
Molecular Weight (g/mol) 
  • (3)
  • (2)
  • (1)
Quantity 
  • (2)
  • (1)
  • (2)
  • (1)
Boiling Point 
  • (1)
  • (2)
Percent Purity 
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)