accessibility menu, dialog, popup

UserName

PdPP

CAS RN 33309-88-5

IUPAC Name: bis(tricyclohexylphosphane) palladium
Synonyms: bis(tricyclohexylphosphine) palladium
Molecular Weight (g/mol): 667.29
Molecular Formula: C36H66P2Pd
InChi Key: JGBZTJWQMWZVNX-UHFFFAOYSA-N
SMILES: [Pd].C1CCC(CC1)P(C1CCCCC1)C1CCCCC1.C1CCC(CC1)P(C1CCCCC1)C1CCCCC1
Physical Form 
  • (2)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)