accessibility menu, dialog, popup

UserName

H3CSOOON

CAS RN 33301-41-6

IUPAC Name: 1-(diphenylmethyl)azetidin-3-yl methanesulfonate
Synonyms: 1-(diphenylmethyl)azetidin-3-yl methanesulfonate
Molecular Weight (g/mol): 317.4
Molecular Formula: C17H19NO3S
InChi Key: MSVZMUILYMLJCF-UHFFFAOYSA-N
SMILES: CS(=O)(=O)OC1CN(C1)C(C1=CC=CC=C1)C1=CC=CC=C1
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results