CAS RN 33252-64-1
CAS RN 33252-64-1
IUPAC Name:
3-nitro-5-(trifluoromethyl)-1,2-dihydropyridin-2-one
Synonyms:
3-nitro-5-(trifluoromethyl)-1H-pyridin-2-one
Molecular Weight (g/mol):
208.1
Molecular Formula:
C6H3F3N2O3
InChi Key:
JYXKHKBZLLIWEV-UHFFFAOYSA-N
SMILES:
[O-][N+](=O)C1=CC(=CNC1=O)C(F)(F)F
1
–
2
of
2
results
1
3-Nitro-5-(trifluoromethyl)pyridin-2-ol, 97%, Thermo Scientific™
CAS: 33252-64-1 Molecular Formula: C6H3F3N2O3 Molecular Weight (g/mol): 208.096 MDL Number: MFCD00153193 InChI Key: JYXKHKBZLLIWEV-UHFFFAOYSA-N PubChem CID: 2775093 IUPAC Name: 3-nitro-5-(trifluoromethyl)-1H-pyridin-2-one SMILES: C1=C(C(=O)NC=C1C(F)(F)F)[N+](=O)[O-]
2
2-Hydroxy-3-nitro-5-(trifluoromethyl)pyridine, 97%, Thermo Scientific™
CAS: 33252-64-1 Molecular Formula: C6H3F3N2O3 Molecular Weight (g/mol): 208.096 MDL Number: MFCD00153193 InChI Key: JYXKHKBZLLIWEV-UHFFFAOYSA-N PubChem CID: 2775093 IUPAC Name: 3-nitro-5-(trifluoromethyl)-1H-pyridin-2-one SMILES: C1=C(C(=O)NC=C1C(F)(F)F)[N+](=O)[O-]