CAS RN 33225-74-0
IUPAC Name:
2-nitropyridine-4-carboxylic acid
Synonyms:
2-nitropyridine-4-carboxylic acid
Molecular Weight (g/mol):
168.11
Molecular Formula:
C6H4N2O4
InChi Key:
SXERXZLYZFAKBX-UHFFFAOYSA-N
SMILES:
OC(=O)C1=CC(=NC=C1)[N+]([O-])=O
1
–
1
of
1
results
1
2-Nitropyridine-4-carboxylic acid, 95%, Thermo Scientific Chemicals
CAS: 33225-74-0 Molecular Formula: C6H4N2O4 Molecular Weight (g/mol): 168.108 MDL Number: MFCD04114160 InChI Key: SXERXZLYZFAKBX-UHFFFAOYSA-N Synonym: 2-nitropyridine-4-carboxylic acid,2-nitroisonicotinic acid,2-nitro-4-pyridinecarboxylic acid,2-nitro-4-pyridine carboxylic acid,2-nitro-isonicotinic acid,2-nitropyridine-4-carboxylicacid,4-pyridinecarboxylic acid, 2-nitro,2-nitroisonicotinicacid,ksc222a1b PubChem CID: 2762837 SMILES: C1=CN=C(C=C1C(=O)O)[N+](=O)[O-]