CAS RN 3322-93-8
CAS RN 3322-93-8
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
1,2-Dibromo-4-(1,2-dibromoethyl)cyclohexane, TRC
CAS: 3322-93-8 Molecular Formula: C8 H12 Br4 Molecular Weight (g/mol): 427.8 Synonym: 1,2-Dibromo-4-(1,2-dibromoethyl)cyclohexane,1-(1,2-Dibromoethyl)-3,4-dibromocyclohexane,4-(1,2-Dibromoethyl)-1,2-dibromocyclohexane,Citex BCL 462,Saytex BCL 462 IUPAC Name: 1,2-dibromo-4-(1,2-dibromoethyl)cyclohexane SMILES: BrCC(Br)C1CCC(Br)C(Br)C1