accessibility menu, dialog, popup

UserName

ONONCO

CAS RN 3320-87-4

IUPAC Name: 1-isocyanato-3-nitrobenzene
Synonyms: benzene, 1-isocyanato-3-nitro-
Molecular Weight (g/mol): 164.12
Molecular Formula: C7H4N2O3
InChi Key: GFFGYTMCNVMFAJ-UHFFFAOYSA-N
SMILES: [O-][N+](=O)C1=CC=CC(=C1)N=C=O
Boiling Point 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)