CAS RN 332-48-9
IUPAC Name:
1-(2-bromoethoxy)-4-fluorobenzene
Synonyms:
1-(2-bromoethoxy)-4-fluorobenzene
Molecular Weight (g/mol):
219.05
Molecular Formula:
C8H8BrFO
InChi Key:
JXSPKRUNMHMICQ-UHFFFAOYSA-N
SMILES:
FC1=CC=C(OCCBr)C=C1
1
–
2
of
2
results
1
1-(2-Bromoethoxy)-4-fluorobenzene, 97+%
CAS: 332-48-9 Molecular Formula: C8H8BrFO Molecular Weight (g/mol): 219.05 MDL Number: MFCD00044739 InChI Key: JXSPKRUNMHMICQ-UHFFFAOYSA-N Synonym: 1-2-bromoethoxy-4-fluorobenzene,4-fluorophenoxy-ethylbromide,2-bromoethyl 4-fluorophenyl ether,4-fluorophenoxyethylbromide,4-2-bromoethoxy fluorobenzene,4-fluorophenoxyethyl bromide,beta-bromo-4-fluorophenetole,2-4-fluorophenoxy ethyl bromide,benzene, 1-2-bromoethoxy-4-fluoro,p-fluorophenoxy ethylbromide PubChem CID: 2064171 IUPAC Name: 1-(2-bromoethoxy)-4-fluorobenzene SMILES: FC1=CC=C(OCCBr)C=C1