accessibility menu, dialog, popup

UserName

H3COBrF

CAS RN 331-61-3

IUPAC Name: 4-(bromomethyl)-2-fluoro-1-methoxybenzene
Synonyms: 4-(bromomethyl)-2-fluoro-1-methoxybenzene
Molecular Weight (g/mol): 219.05
Molecular Formula: C8H8BrFO
InChi Key: KLWYYWVSFYRPLM-UHFFFAOYSA-N
SMILES: COC1=CC=C(CBr)C=C1F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results