accessibility menu, dialog, popup

UserName

NNFFFCl

CAS RN 33034-67-2

IUPAC Name: 2-chloro-4-(trifluoromethyl)pyrimidine
Synonyms: 2-chloro-4-(trifluoromethyl)pyrimidine
Molecular Weight (g/mol): 182.53
Molecular Formula: C5H2ClF3N2
InChi Key: FZRBTBCCMVNZBD-UHFFFAOYSA-N
SMILES: FC(F)(F)C1=NC(Cl)=NC=C1
Quantity 
  • (3)
  • (1)
  • (3)
Boiling Point 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (3)
  • (1)
  • (3)
  • (2)
  • (2)
  • (2)