accessibility menu, dialog, popup

UserName

H3CNCH3ONHClCl

CAS RN 330-54-1

IUPAC Name: 1-(3,4-dichlorophenyl)-3,3-dimethylurea
Synonyms: dynex marmer diuron (iso) diuron DCMU
Molecular Weight (g/mol): 233.09
Molecular Formula: C9H10Cl2N2O
InChi Key: XMTQQYYKAHVGBJ-UHFFFAOYSA-N
SMILES: CN(C)C(=O)NC1=CC=C(Cl)C(Cl)=C1
Molecular Weight (g/mol) 
  • (2)
  • (1)
  • (9)
Solvent or Matrix 
  • (2)
  • (2)
  • (3)
Concentration 
  • (3)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
Boiling Point 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (6)
  • (1)
  • (1)
  • (2)
  • (1)
  • (9)
  • (2)
  • (2)
  • (3)
  • (3)
  • (3)
  • (1)
  • (1)
  • (1)
  • (2)