accessibility menu, dialog, popup

UserName

H3CNHONHNCH3CH3

CAS RN 32897-26-0

IUPAC Name: 3-[3-(dimethylamino)propyl]-1-ethylurea
Synonyms: 3-[3-(dimethylamino)propyl]-1-ethylurea
Molecular Weight (g/mol): 173.26
Molecular Formula: C8H19N3O
InChi Key: NGJUYARYEXGDNN-UHFFFAOYSA-N
SMILES: CCNC(=O)NCCCN(C)C
Quantity 
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results