accessibility menu, dialog, popup

UserName

CAS RN 328-74-5

IUPAC Name: 3,5-bis(trifluoromethyl)aniline
Synonyms: 3,5-bis(trifluorome)aniline
Molecular Weight (g/mol): 229.13
Molecular Formula: C8H5F6N
InChi Key: CDIDGWDGQGVCIB-UHFFFAOYSA-N
SMILES: NC1=CC(=CC(=C1)C(F)(F)F)C(F)(F)F
Boiling Point 
  • (3)
  • (3)
Molecular Weight (g/mol) 
  • (3)
  • (3)
Percent Purity 
  • (3)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (3)
  • (3)
  • (3)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)