accessibility menu, dialog, popup

UserName

ClNBrN

CAS RN 32779-36-5

IUPAC Name: 5-bromo-2-chloropyrimidine
Synonyms: 5-bromo-2-chloropyrimidine
Molecular Weight (g/mol): 193.43
Molecular Formula: C4H2BrClN2
InChi Key: XPGIBDJXEVAVTO-UHFFFAOYSA-N
SMILES: ClC1=NC=C(Br)C=N1
Quantity 
  • (2)
  • (1)
  • (1)
  • (3)
Percent Purity 
  • (3)
  • (3)
Boiling Point 
  • (3)
Physical Form 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (3)
  • (3)
  • (3)
  • (3)
  • (3)