CAS RN 32778-09-9
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
2
of
2
results
1
1,1-Dichloro-1-(chlorodifluoromethoxy)-2,2,2-trifluoroethane, TRC
CAS: 32778-09-9 Molecular Formula: C3 Cl3 F5 O Molecular Weight (g/mol): 253.38 Synonym: 1,1-Dichloro-1-(chlorodifluoromethoxy)-2,2,2-trifluoroethane,Isoflurane Imp. E (EP) IUPAC Name: 1,1-dichloro-1-[chloro(difluoro)methoxy]-2,2,2-trifluoroethane SMILES: FC(F)(F)C(Cl)(Cl)OC(F)(F)Cl
2
1,1-Dichloro-1-(chlorodifluoromethoxy)-2,2,2-trifluoroethane, Mikromol™
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.