accessibility menu, dialog, popup

UserName

SHOOOHOOHNHNH2N(S)(R)(R)CH3CH3(R)H

CAS RN 32746-94-4

IUPAC Name:
Synonyms:
Molecular Weight (g/mol):
Molecular Formula:
InChi Key:
SMILES:

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)