accessibility menu, dialog, popup

UserName

CAS RN 327056-73-5

FClN

CAS RN 327056-73-5

IUPAC Name: 3-chloro-5-fluorobenzonitrile
Synonyms: 3-chloro-5-fluorobenzonitrile
Molecular Weight (g/mol): 155.56
Molecular Formula: C7H3ClFN
InChi Key: GIKLRWNRBOLRDV-UHFFFAOYSA-N
SMILES: FC1=CC(Cl)=CC(=C1)C#N
Molecular Weight (g/mol) 
  • (2)
  • (3)
Percent Purity 
  • (2)
  • (3)
Quantity 
  • (1)
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (2)
  • (3)
  • (2)
  • (3)
  • (1)
  • (2)
  • (2)