accessibility menu, dialog, popup

UserName

ONOHN

CAS RN 32692-19-6

IUPAC Name: 5-nitro-2,3-dihydro-1H-indole
Synonyms: 5-nitro-2,3-dihydro-1H-indole
Molecular Weight (g/mol): 164.16
Molecular Formula: C8H8N2O2
InChi Key: WJQWYAJTPPYORB-UHFFFAOYSA-N
SMILES: [O-][N+](=O)C1=CC=C2NCCC2=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results