CAS RN 32679-03-1
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Bis(acetonitrile)(1,5-cyclooctadiene)iridium(I) tetrafluoroborate, 97%
CAS: 32679-03-1 Molecular Formula: C12H18BF4IrN2 Molecular Weight (g/mol): 469.31 MDL Number: MFCD28144566 InChI Key: VEAARGXUKIPMQB-SUESZSCISA-N IUPAC Name: iridium(1+) (1Z,5Z)-cycloocta-1,5-diene bis(acetonitrile) tetrafluoroboranuide SMILES: [Ir+].CC#N.CC#N.F[B-](F)(F)F.C1C\C=C/CC\C=C/1