accessibility menu, dialog, popup

UserName

CAS RN 32587-10-3

H2NONNH2N

CAS RN 32587-10-3

IUPAC Name: 3-aminopyrazine-2-carboxamide
Synonyms: 3-aminopyrazine-2-carboxamide
Molecular Weight (g/mol): 138.13
Molecular Formula: C5H6N4O
InChi Key: SFSMATGDLFHTHE-UHFFFAOYSA-N
SMILES: NC(=O)C1=NC=CN=C1N
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results