CAS RN 32510-61-5
CAS RN 32510-61-5
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Cefazolin 3-Acetyloxy, TRC
CAS: 32510-61-5 Molecular Formula: C13H14N6O6S Molecular Weight (g/mol): 382.35 Synonym: (6R,7R)-3-[(Acetyloxy)methyl]-8-oxo-7-[[2-(1H-tetrazol-1-yl)acetyl]amino]-5-thia-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylic acid,7-(1H-Tetrazol-1-acetamido)cephalosporanic acid,N-(Tetrazolylacetyl)-7-aminocephalosporin IUPAC Name: (6R,7R)-3-(acetyloxymethyl)-8-oxo-7-[[2-(tetrazol-1-yl)acetyl]amino]-5-thia-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylic acid SMILES: CC(=O)OCC1=C(N2[C@H](SC1)[C@H](NC(=O)Cn3cnnn3)C2=O)C(=O)O