accessibility menu, dialog, popup

UserName

CAS RN 32449-92-6

OH(R)O(S)OO(S)HO(R)HO

CAS RN 32449-92-6

IUPAC Name: (2R)-2-[(2S,3R,4S)-3,4-dihydroxy-5-oxooxolan-2-yl]-2-hydroxyacetaldehyde
Synonyms: glucuronolactone D-glucuronic acid lactone D-glucurono-3,6-lactone glucurolactone D-glucurono-6,3-lactone
Molecular Weight (g/mol): 176.12
Molecular Formula: C6H8O6
InChi Key: UYUXSRADSPPKRZ-SKNVOMKLSA-N
SMILES: O[C@@H](C=O)[C@H]1OC(=O)[C@@H](O)[C@H]1O
Molecular Weight (g/mol) 
  • (2)
  • (4)
Percent Purity 
  • (1)
  • (3)
  • (2)
Quantity 
  • (2)
  • (2)
  • (2)
Physical Form 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (1)
  • (2)
  • (4)
  • (1)
  • (3)
  • (2)
  • (2)
  • (2)
  • (2)
  • (3)