accessibility menu, dialog, popup

UserName

CAS RN 3226-34-4

H3COClOH

CAS RN 3226-34-4

IUPAC Name: 1-(3-chloro-2-hydroxyphenyl)ethan-1-one
Synonyms: 1-(3-chloro-2-hydroxyphenyl)ethanone
Molecular Weight (g/mol): 170.59
Molecular Formula: C8H7ClO2
InChi Key: GBWVDQBTXFIIJF-UHFFFAOYSA-N
SMILES: CC(=O)C1=C(O)C(Cl)=CC=C1
Grade 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results