accessibility menu, dialog, popup

UserName

CAS RN 32246-20-1

H3COOHOBrOCH3

CAS RN 32246-20-1

IUPAC Name: 5-bromo-2,4-dimethoxybenzoic acid
Synonyms: 5-bromo-2,4-dimethoxybenzoic acid
Molecular Weight (g/mol): 261.07
Molecular Formula: C9H9BrO4
InChi Key: WOPJFSQYDOHZMK-UHFFFAOYSA-N
SMILES: COC1=CC(OC)=C(Br)C=C1C(O)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results