CAS RN 321705-77-5
IUPAC Name:
3,4-difluoro-N-[(4-methylphenyl)methyl]benzene-1-sulfonamide
Synonyms:
3,4-difluoro-N-[(4-methylphenyl)methyl]benzenesulfonamide
Molecular Weight (g/mol):
297.32
Molecular Formula:
C14H13F2NO2S
InChi Key:
LUPVVMZLWZQPGV-UHFFFAOYSA-N
SMILES:
CC1=CC=C(CNS(=O)(=O)C2=CC(F)=C(F)C=C2)C=C1
1
–
1
of
1
results
1
3,4-Difluoro-N-(4-methylbenzyl)benzenesulfonamide, 97%, Thermo Scientific™
CAS: 321705-77-5 Molecular Formula: C14H13F2NO2S Molecular Weight (g/mol): 297.32 MDL Number: MFCD02556071 InChI Key: LUPVVMZLWZQPGV-UHFFFAOYSA-N Synonym: 3,4-difluoro-n-4-methylphenyl methyl benzenesulfonamide,3,4-difluoro-n-4-methylbenzyl benzenesulfonamide,3,4-difluorophenyl sulfonyl 4-methylphenyl methyl amine,3,4-difluoro-n-4-methylphenyl methyl benzene-1-sulfonamide PubChem CID: 6467917 IUPAC Name: 3,4-difluoro-N-[(4-methylphenyl)methyl]benzenesulfonamide SMILES: CC1=CC=C(CNS(=O)(=O)C2=CC(F)=C(F)C=C2)C=C1