accessibility menu, dialog, popup

UserName

NS

CAS RN 3216-48-6

IUPAC Name: 2-(1-benzothiophen-3-yl)acetonitrile
Synonyms: 2-(1-benzothiophen-3-yl)acetonitrile
Molecular Weight (g/mol): 173.23
Molecular Formula: C10H7NS
InChi Key: DEELHLMCCWZFFE-UHFFFAOYSA-N
SMILES: N#CCC1=CSC2=CC=CC=C12

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results