accessibility menu, dialog, popup

UserName

CH3H3CNOClNOO

CAS RN 321531-52-6

IUPAC Name: N-benzyl-4-chloro-3-nitro-N-(propan-2-yl)benzamide
Synonyms: N-benzyl-4-chloro-N-isopropyl-3-nitrobenzamide
Molecular Weight (g/mol): 332.78
Molecular Formula: C17H17ClN2O3
InChi Key: NGZSJSBJVCIBDV-UHFFFAOYSA-N
SMILES: CC(C)N(CC1=CC=CC=C1)C(=O)C1=CC(=C(Cl)C=C1)[N+]([O-])=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results