accessibility menu, dialog, popup

UserName

CAS RN 32149-57-8

MgH3CCH3CH3O

CAS RN 32149-57-8

IUPAC Name: magnesium(2+) 2-methylpropan-2-olate
Synonyms: magnesium(2+) 2-methylpropan-2-olate
Molecular Weight (g/mol): 97.42
Molecular Formula: C4H9MgO
InChi Key: ONFWJTUNDMIGEP-UHFFFAOYSA-N
SMILES: [Mg++].CC(C)(C)[O-]
Molecular Weight (g/mol) 
  • (3)
  • (3)
  • (3)
Quantity 
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
Percent Purity 
  • (3)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (3)
  • (3)
  • (3)
  • (3)
  • (3)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (3)
  • (3)
  • (3)