accessibility menu, dialog, popup

UserName

CAS RN 321309-40-4

SSOOClNO

CAS RN 321309-40-4

IUPAC Name: 5-(1,3-oxazol-5-yl)thiophene-2-sulfonyl chloride
Synonyms: 5-(1,3-oxazol-5-yl)thiophene-2-sulfonyl chloride
Molecular Weight (g/mol): 249.68
Molecular Formula: C7H4ClNO3S2
InChi Key: MAGTZUMWQUDWAH-UHFFFAOYSA-N
SMILES: ClS(=O)(=O)C1=CC=C(S1)C1=CN=CO1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results