CAS RN 32113-41-0
CAS RN 32113-41-0
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
4-Acetaminophen Sulfate Potassium Salt, TRC
CAS: 32113-41-0 Molecular Formula: C8 H8 N O5 S . K Molecular Weight (g/mol): 269.32 Synonym: Acetamide, N-[4-(sulfooxy)phenyl]-, potassium salt (1:1) (ACI),Acetamide, N-[4-(sulfooxy)phenyl]-, monopotassium salt (9CI),Acetanilide, 4'-hydroxy-, hydrogen sulfate (ester), monopotassium salt (8CI),Potassium N-acetyl-p-aminophenyl sulfate,N-Acetyl-p-aminophenyl sulfate potassium salt,Paracetamol O-sulfate potassium salt,Paracetamol sulfate potassium,Potassium paracetamol O-sulfate,Paracetamol O-sulfate potassium IUPAC Name: potassium;(4-acetamidophenyl) sulfate SMILES: [K+].CC(=O)Nc1ccc(OS(=O)(=O)[O-])cc1