accessibility menu, dialog, popup

UserName

CAS RN 32111-21-0

INN

CAS RN 32111-21-0

IUPAC Name: 2-iodopyrazine
Synonyms: 2-iodopyrazine
Molecular Weight (g/mol): 205.99
Molecular Formula: C4H3IN2
InChi Key: OYWPFIUVDKHHGQ-UHFFFAOYSA-N
SMILES: IC1=CN=CC=N1
Boiling Point 
  • (2)
  • (2)
Molecular Weight (g/mol) 
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)