accessibility menu, dialog, popup

UserName

CAS RN 321-07-3

IUPAC Name: 1-phenyl-3-(trifluoromethyl)-4,5-dihydro-1H-pyrazol-5-one
Synonyms: 2-phenyl-5-(trifluoromethyl)-4H-pyrazol-3-one
Molecular Weight (g/mol): 228.17
Molecular Formula: C10H7F3N2O
InChi Key: GLGRRRKQSFURGD-UHFFFAOYSA-N
SMILES: FC(F)(F)C1=NN(C(=O)C1)C1=CC=CC=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (1)
  • (1)