accessibility menu, dialog, popup

UserName

FFFClO

CAS RN 320-43-4

IUPAC Name: 4-chloro-2-(trifluoromethyl)benzaldehyde
Synonyms: 4-chloro-2-(trifluoromethyl)benzaldehyde
Molecular Weight (g/mol): 208.56
Molecular Formula: C8H4ClF3O
InChi Key: AHFINSWGYAZBOZ-UHFFFAOYSA-N
SMILES: FC(F)(F)C1=CC(Cl)=CC=C1C=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results