accessibility menu, dialog, popup

UserName

ONO

CAS RN 31909-58-7

IUPAC Name: 3-(furan-2-yl)-3-oxopropanenitrile
Synonyms: 3-(furan-2-yl)-3-oxopropanenitrile
Molecular Weight (g/mol): 135.12
Molecular Formula: C7H5NO2
InChi Key: RZNSHBXVTAHWPP-UHFFFAOYSA-N
SMILES: O=C(CC#N)C1=CC=CO1
Percent Purity 
  • (3)
  • (1)
Quantity 
  • (2)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results