CAS RN 31883-05-3
CAS RN 31883-05-3
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Moricizine, TRC
CAS: 31883-05-3 Molecular Formula: C22H25N3O4S Molecular Weight (g/mol): 427.52 Synonym: N-[10-[3-(4-morpholinyl)-1-oxopropyl]-10H-phenothiazin-2-yl]Carbamic Acid Ethyl Ester,10-(3-Morpholinopropionyl)phenothiazine-2-carbamic Acid Ethyl Ester,Ethyl 10-(beta-N-Morpholinylpropionyl)phenothiazine-2-carbamate,Ethyl 10-(beta-Morpholinopropionyl)phenothiazine-2-carbamate,G 214,Moracizine IUPAC Name: ethyl N-[10-(3-morpholin-4-ylpropanoyl)phenothiazin-2-yl]carbamate SMILES: CCOC(=O)Nc1ccc2Sc3ccccc3N(C(=O)CCN4CCOCC4)c2c1