accessibility menu, dialog, popup

UserName

CAS RN 3185-99-7

CH3SCH3OO

CAS RN 3185-99-7

IUPAC Name: 1-methanesulfonyl-4-methylbenzene
Synonyms: P-methylsulfonyltoluene
Molecular Weight (g/mol): 170.23
Molecular Formula: C8H10O2S
InChi Key: YYDNBUBMBZRNQQ-UHFFFAOYSA-N
SMILES: CC1=CC=C(C=C1)S(C)(=O)=O
Boiling Point 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results