accessibility menu, dialog, popup

UserName

CH3OO(S)NH2HCl

CAS RN 3182-93-2

IUPAC Name: hydrogen ethyl (2S)-2-amino-3-phenylpropanoate chloride
Synonyms: hydrogen ethyl (2S)-2-amino-3-phenylpropanoate chloride
Molecular Weight (g/mol): 229.7
Molecular Formula: C11H16ClNO2
InChi Key: FPFQPLFYTKMCHN-PPHPATTJSA-N
SMILES: [H+].[Cl-].CCOC(=O)[C@@H](N)CC1=CC=CC=C1
Percent Purity 
  • (3)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (3)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)