accessibility menu, dialog, popup

UserName

CH3OOHNFO

CAS RN 318-35-4

IUPAC Name: ethyl 6-fluoro-4-oxo-1,4-dihydroquinoline-3-carboxylate
Synonyms: ethyl 6-fluoro-4-oxo-1H-quinoline-3-carboxylate
Molecular Weight (g/mol): 235.21
Molecular Formula: C12H10FNO3
InChi Key: LNPRCADPRULVTR-UHFFFAOYSA-N
SMILES: CCOC(=O)C1=CNC2=CC=C(F)C=C2C1=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results