accessibility menu, dialog, popup

UserName

H3COBrOOH

CAS RN 31786-45-5

IUPAC Name: 2-bromo-6-methoxybenzoic acid
Synonyms: 2-bromo-6-methoxybenzoic acid
Molecular Weight (g/mol): 231.05
Molecular Formula: C8H7BrO3
InChi Key: BNYFDICJYCBPRW-UHFFFAOYSA-N
SMILES: COC1=C(C(O)=O)C(Br)=CC=C1
Molecular Weight (g/mol) 
  • (3)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (3)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)