accessibility menu, dialog, popup

UserName

CAS RN 3173-56-6

OCN

CAS RN 3173-56-6

IUPAC Name: (isocyanatomethyl)benzene
Synonyms: (isocyanatomethyl)benzene
Molecular Weight (g/mol): 133.15
Molecular Formula: C8H7NO
InChi Key: YDNLNVZZTACNJX-UHFFFAOYSA-N
SMILES: O=C=NCC1=CC=CC=C1
Quantity 
  • (1)
  • (3)
  • (2)
Boiling Point 
  • (2)
  • (3)
Percent Purity 
  • (3)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (1)
  • (1)
  • (3)
  • (2)
  • (2)
  • (3)
  • (3)
  • (2)
  • (2)