accessibility menu, dialog, popup

UserName

OCN

CAS RN 3173-53-3

IUPAC Name: isocyanatocyclohexane
Synonyms: cyclohexyl isocyanate
Molecular Weight (g/mol): 125.17
Molecular Formula: C7H11NO
InChi Key: KQWGXHWJMSMDJJ-UHFFFAOYSA-N
SMILES: O=C=NC1CCCCC1
Percent Purity 
  • (1)
  • (1)
Quantity 
  • (1)
  • (1)
Boiling Point 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)