accessibility menu, dialog, popup

UserName

CAS RN 31719-75-2

OOOH

CAS RN 31719-75-2

IUPAC Name: 3-(phenoxymethyl)benzoic acid
Synonyms: 3-(phenoxymethyl)benzoic acid
Molecular Weight (g/mol): 228.25
Molecular Formula: C14H12O3
InChi Key: URLYREZIPSJJQU-UHFFFAOYSA-N
SMILES: OC(=O)C1=CC(COC2=CC=CC=C2)=CC=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results